accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1676104-71-4

OOOOOOOOOOHOOONHHN(R)(R)(R)(R)(R)(S)CH3CH3H3CCH3

CAS RN 1676104-71-4

IUPAC Name: (2S)-3-{[(2R,3R,4R,5R,6R)-4,5-bis(acetyloxy)-6-[(acetyloxy)methyl]-3-acetamidooxan-2-yl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-{[(2R,3R,4R,5R,6R)-4,5-bis(acetyloxy)-6-[(acetyloxy)methyl]-3-acetamidooxan-2-yl]oxy}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 656.64
Molecular Formula: C32H36N2O13
InChi Key: ORICVOOXZDVFIP-UKFUGAQESA-N
SMILES: CC(=O)N[C@H]1[C@H](OC[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)O[C@H](COC(C)=O)[C@H](OC(C)=O)[C@@H]1OC(C)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)