accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 167683-93-4

IUPAC Name: 2-fluoro-4-methoxyphenol
Synonyms: 2-fluoro-4-methoxyphenol
Molecular Weight (g/mol): 142.13
Molecular Formula: C7H7FO2
InChi Key: FCJHBXCQIOVMEM-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results