Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-methoxyphenol 95.0+%, TCI America™
SDP

Catalog No. F06345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06345G Supplier TCI America Supplier No. F06345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 167683-93-4
Molecular Formula C7H7FO2
Molecular Weight (g/mol) 142.129
MDL Number MFCD00070796
InChI Key FCJHBXCQIOVMEM-UHFFFAOYSA-N
Synonym 3-Fluoro-4-hydroxyanisole
PubChem CID 2774557
IUPAC Name 2-fluoro-4-methoxyphenol
SMILES COC1=CC(=C(C=C1)O)F

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 142.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-4-methoxyphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.