accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOONNHSOOH3CClNH2OCH3HCl

CAS RN 167710-87-4

IUPAC Name: N-{2-[4-(3-{4-amino-5-chloro-2-[(3,5-dimethoxyphenyl)methoxy]phenyl}-3-oxopropyl)piperidin-1-yl]ethyl}methanesulfonamide hydrochloride
Synonyms: RS 39604 hydrochloride
Molecular Weight (g/mol): 590.56
Molecular Formula: C26H37Cl2N3O6S
InChi Key: QSMYZGMJSGUWPM-UHFFFAOYSA-N
SMILES: Cl.COC1=CC(OC)=CC(COC2=CC(N)=C(Cl)C=C2C(=O)CCC2CCN(CCNS(C)(=O)=O)CC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results