CAS RN 16774-21-3
Ammonium cerium(IV) nitrate, 99%, for analysis
CAS: 16774-21-3 Molecular Formula: H8CeN8O18 Molecular Weight (g/mol): 548.22 MDL Number: MFCD00151121 Synonym: Ceric ammonium nitrate,CAN
Ammonium cerium(IV) nitrate, pure
CAS: 16774-21-3 Molecular Formula: H8CeN8O18 Molecular Weight (g/mol): 548.22 Synonym: Ceric ammonium nitrate,CAN
Ammonium cerium(IV) nitrate, 99%, ACS reagent
CAS: 16774-21-3 Molecular Formula: H8CeN8O18 Molecular Weight (g/mol): 548.22 MDL Number: MFCD00151121 Synonym: Ceric ammonium nitrate,CAN
Cerium(IV) ammonium nitrate, ACS, 98.5% min
CAS: 16774-21-3 Molecular Formula: (NH4)2Ce(NO3)6 MDL Number: MFCD00151121
Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO)
CAS: 16774-21-3 Molecular Formula: H8CeN8O18 MDL Number: MFCD00151121
Cerium(IV) ammonium nitrate, 98+%
CAS: 16774-21-3 Molecular Formula: H8CeN8O18 MDL Number: MFCD00151121 Synonym: Ammonium cerium(IV) nitrate; CAN
Ceric Ammonium Sulfate, in 1.0N Sulfuric Acid, Certified, 0.100N ±0.001N (0.10M), LabChem™
CAS: 16774-21-3 Molecular Formula: CeH8N8O18 Molecular Weight (g/mol): 548.22 InChI Key: WIBGOERAEYJBOT-UHFFFAOYSA-N Synonym: Ceric ammonium nitrate,CAN PubChem CID: 24181046 IUPAC Name: λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate SMILES: N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Ammonium Cerium(IV) Nitrate 98.0+%, TCI America™
CAS: 16774-21-3 Molecular Formula: H8CeN8O18 MDL Number: MFCD00151121 Synonym: Ceric Ammonium Nitrate, CAN, Cerium(IV) Diammonium Nitrate, Diammonium Cerium(IV) Nitrate
Ceric Ammonium Nitrate, ACS Reagent Grade, Ricca Chemical
CAS: 16774-21-3 Molecular Weight (g/mol): 548.26 g/mol
Ceric Ammonium Nitrate, ACS Reagent (Ammonium Cerium (IV) Nitrate), Reagents
CAS: 16774-21-3 Molecular Formula: CeH8N8O18 Molecular Weight (g/mol): 548.22 InChI Key: WIBGOERAEYJBOT-UHFFFAOYSA-N Synonym: Ammonium Cerium(IV) Nitrate, Ammonium Hexanitratocerate (IV) IUPAC Name: λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate SMILES: N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Ammonium cerium(IV) nitrate solution, Solstice
CAS: 16774-21-3 Molecular Weight (g/mol): 548.23 MDL Number: MFCD00151121
Sigma Aldrich Ammonium cerium(IV) nitrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Ceric Ammonium Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 16774-21-3 Molecular Formula: (NH4)2Ce(NO3)6
Ammonium Cerium(Iv) Nitrate, ≥98.0% (T), purum p.a., Solstice
CAS: 16774-21-3 MDL Number: MFCD00151121
- 1
- 2