Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO)

Catalog No. AA1133236
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1133236 500 g
AA1133209 10 g
AA1133222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1133236 Supplier Thermo Scientific Chemicals Supplier No. 01133236
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16774-21-3 | H8CeN8O18

Cerium(IV) ammonium nitrate is used to offers a novel route to tartronic acid derivatives via facile oxygenation of malonates. It is also used as for oxidative addition reactions of electrophilic radicals to alkenes, enabling intermolecular and intramolecular carbon-carbon and carbon-heteroatom bond formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
MDL Number MFCD00151121

Specifications

Color Yellow to Orange
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.5% (REO)
Solubility Information Soluble in water.
Formula Weight 548.23
Odor Pungent
Chemical Name or Material Cerium(IV) ammonium nitrate
Hazard Category H272-H290-H302-H314-H317-H335
Precautionary Statement P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 240-827-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.