CAS RN 1680193-80-9
IUPAC Name:
13-chloro-2-[1-(1H-1,2,4-triazole-1-carbonyl)piperidin-4-ylidene]-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(11),3(8),4,6,12,14-hexaene
Synonyms:
13-chloro-2-[1-(1,2,4-triazole-1-carbonyl)piperidin-4-ylidene]-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(11),3(8),4,6,12,14-hexaene
Molecular Weight (g/mol):
405.89
Molecular Formula:
C22H20ClN5O
InChi Key:
GAVZCGTYRWKKDV-UHFFFAOYSA-N
SMILES:
ClC1=CC2=C(C=C1)C(=C1CCN(CC1)C(=O)N1C=NC=N1)C1=C(CC2)C=CC=N1
1
–
1
of
1
results
1
Tocris Bioscience™ JZP 361, >99%
Potent and selective reversible MAGL inhibitor