CAS RN 168982-69-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(3-Oxododecanoyl)-L-homoserine lactone, Tocris Bioscience™
Immunosuppressants