accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169209-63-6

IUPAC Name: 4-aminopyrrolidine-2,4-dicarboxylic acid
Synonyms: 4-aminopyrrolidine-2,4-dicarboxylic acid
Molecular Weight (g/mol): 174.16
Molecular Formula: C6H10N2O4
InChi Key: XZFMJVJDSYRWDQ-UHFFFAOYNA-N
SMILES: NC1(CNC(C1)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)