accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16939-73-4

IUPAC Name: (2E)-hept-2-en-1-yl acetate
Synonyms: (2E)-hept-2-en-1-yl acetate
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: AWCPMVVOGVEPRC-VOTSOKGWSA-N
SMILES: CCCC\C=C\COC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results