CAS RN 169505-93-5
IUPAC Name:
[1-(5-chloro-1H-indol-3-yl)-2-methylpropan-2-yl]({2-[2-(cyclopropylmethoxy)phenoxy]ethyl})amine hydrochloride
Synonyms:
[1-(5-chloro-1H-indol-3-yl)-2-methylpropan-2-yl]({2-[2-(cyclopropylmethoxy)phenoxy]ethyl})amine hydrochloride
Molecular Weight (g/mol):
449.42
Molecular Formula:
C24H30Cl2N2O2
InChi Key:
QFOPFGRPNPCPBX-UHFFFAOYSA-N
SMILES:
Cl.CC(C)(CC1=CNC2=CC=C(Cl)C=C12)NCCOC1=CC=CC=C1OCC1CC1
1
–
1
of
1
results
1
Tocris Bioscience™ RS 17053 hydrochloride
CAS: 169505-93-5 Molecular Formula: C24H30Cl2N2O2