accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHO(E)

CAS RN 16957-70-3

IUPAC Name: (2E)-2-methylpent-2-enoic acid
Synonyms: (2E)-2-methylpent-2-enoic acid
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: JJYWRQLLQAKNAD-SNAWJCMRSA-N
SMILES: CC\C=C(/C)C(O)=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)