Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Methyl-2-pentenoic acid, 97%, Thermo Scientific™

Catalog No. AAH6039114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6039114 25 g
AAH6039106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6039114 Supplier Thermo Scientific Chemicals Supplier No. H6039114

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16957-70-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00002655
InChI Key JJYWRQLLQAKNAD-SNAWJCMRSA-N
Synonym 2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
PubChem CID 5365909
IUPAC Name (E)-2-methylpent-2-enoic acid
SMILES CCC=C(C)C(=O)O

Specifications

Melting Point 26°C to 28°C
Density 0.979
Boiling Point 123°C to 125°C (30 mmHg)
Flash Point 108°C (226°F)
Refractive Index 1.46
Linear Formula C2H5CH=C(CH3)CO2H
Quantity 25 g
UN Number UN3261
Formula Weight 114.14
Percent Purity 97%
Chemical Name or Material trans-2-Methyl-2-pentenoic acid
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 241-026-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.