Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Methyl-2-pentenoic Acid 97.0+%, TCI America™
SDP

Catalog No. M28145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28145G 5 g
M281425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28145G Supplier TCI America Supplier No. M28145G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16957-70-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00002655
InChI Key JJYWRQLLQAKNAD-SNAWJCMRSA-N
Synonym 2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
PubChem CID 5365909
IUPAC Name (E)-2-methylpent-2-enoic acid
SMILES CCC=C(C)C(=O)O

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
UN Number 3265
Formula Weight 114.14
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material trans-2-Methyl-2-pentenoic Acid
EINECSNumber (2)-0609
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.