accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169896-41-7

IUPAC Name: [bis(diethylamino)(trimethylimino)tantalio]diethylamine
Synonyms: [bis(diethylamino)(trimethylimino)tantalio]diethylamine
Molecular Weight (g/mol): 456.45
Molecular Formula: C15H39N4Ta
InChi Key: NXXDVZOFTYVIKX-UHFFFAOYSA-N
SMILES: CCN(CC)[Ta](N(CC)CC)(N(CC)CC)=[N](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)