Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis)

Catalog No. AA4341914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4341914 25 g
AA4341906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4341914 Supplier Thermo Scientific Chemicals Supplier No. 04341914
Add to Cart

missing translation for 'validMainframeMsg'

Precursor for TaN | CAS: 169896-41-7 | C15H39N4Ta | 456.45 g/mol

Tantalum tris(diethylamido)-tert-butylimide is used as a precursor for tantalum nitride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169896-41-7
Molecular Formula C15H39N4Ta
Molecular Weight (g/mol) 456.45
MDL Number MFCD02684506
InChI Key NXXDVZOFTYVIKX-UHFFFAOYSA-N
Synonym tert-butylimino tris diethylamino tantalum, tantalum tris diethylamido-tert-butylimide, tbtdet, tris diethylamino tert-butylimino tantalum, t-butylimido tris diethylamino tantalum v, tris diethylamido tert-butylimido tantalum v, tris diethylamino tert-butylimino tantalum v, tert-butylimino bis diethylamino tantalio diethylamine, tantalum,tris n-ethylethanaminato 2-methyl-2-propanaminato 2--, t-4
PubChem CID 4100858
IUPAC Name tert-butyliminotantalum;diethylazanide
SMILES CCN(CC)[Ta](N(CC)CC)(N(CC)CC)=[N](C)(C)C

Specifications

Color Yellow
Boiling Point 90°C (0.1 mmHg)
Packaging Ampule
Quantity 25 g
UN Number UN3129
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 468.48
Percent Purity 99.99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tantalum tris (diethylamido)-tert-butylimide
Show More Show Less
Hazard Category H261-H314-H335
Hazard Statement GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
TSCA Yes
storageNote1
  • Ampouled under argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.