accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHH2N

CAS RN 170080-13-4

IUPAC Name: 2-amino-3-{[1,1'-biphenyl]-4-yl}propanoic acid
Synonyms: 2-amino-3-{[1,1'-biphenyl]-4-yl}propanoic acid
Molecular Weight (g/mol): 241.29
Molecular Formula: C15H15NO2
InChi Key: JCZLABDVDPYLRZ-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(C=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)