Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Biphenylyl)-D-alanine, 98%, Thermo Scientific™

Catalog No. AAH63785MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63785MD 250 mg
AAH6378503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63785MD Supplier Thermo Scientific Chemicals Supplier No. H63785MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170080-13-4
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD01318879
InChI Key JCZLABDVDPYLRZ-CQSZACIVSA-N
Synonym r-3-1,1'-biphenyl-4-yl-2-aminopropanoic acid, d-4,4'-biphenylalanine, h-4-phenyl-d-phe-oh, 4-phenyl-d-phenylalanine, d-biphenylalanine, 4-phenyl-d-phenyalanine, d-4,4'-biphenylphenylalanine, h-d-bip-oh, 4'-biphenyl-d-ala, 2r-2-amino-3-4-phenylphenyl propanoic acid
PubChem CID 7006711
IUPAC Name (2R)-2-amino-3-(4-phenylphenyl)propanoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)N

Specifications

Color White
Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 241.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(4-Biphenylyl)-D-alanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.