accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17025-47-7

IUPAC Name: tribromomethanesulfonylbenzene
Synonyms: tribromomethanesulfonylbenzene
Molecular Weight (g/mol): 392.89
Molecular Formula: C7H5Br3O2S
InChi Key: DWWMSEANWMWMCB-UHFFFAOYSA-N
SMILES: BrC(Br)(Br)S(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results