Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribromomethyl Phenyl Sulfone 97.0+%, TCI America™
SDP

Catalog No. T118825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T118825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T118825G Supplier TCI America Supplier No. T118825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17025-47-7
Molecular Formula C7H5Br3O2S
Molecular Weight (g/mol) 392.887
MDL Number MFCD00060068
InChI Key DWWMSEANWMWMCB-UHFFFAOYSA-N
Synonym Phenyl Tribromomethyl Sulfone
PubChem CID 86912
IUPAC Name tribromomethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)C(Br)(Br)Br

Specifications

Melting Point 147°C
Color White
Quantity 25 g
Formula Weight 392.89
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tribromomethyl Phenyl Sulfone
EINECSNumber (3)-3193
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.