accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 170278-50-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-(diphenylphosphanyl)propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3-(diphenylphosphanyl)propanoate
Molecular Weight (g/mol): 355.33
Molecular Formula: C19H18NO4P
InChi Key: QRRPSWGREIRROT-UHFFFAOYSA-N
SMILES: O=C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results