CAS RN 170364-57-5
IUPAC Name:
3-(1-methyl-1H-indol-3-yl)-4-(1-{1-[(pyridin-2-yl)methyl]piperidin-4-yl}-1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms:
enzastaurin
Molecular Weight (g/mol):
515.62
Molecular Formula:
C32H29N5O2
InChi Key:
AXRCEOKUDYDWLF-UHFFFAOYSA-N
SMILES:
CN1C=C(C2=CC=CC=C12)C1=C(C(=O)NC1=O)C1=CN(C2CCN(CC3=CC=CC=N3)CC2)C2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Enzastaurin, 99%
Thermo Scientific Chemicals Enzastaurin, 99%
CAS: 170364-57-5 Molecular Formula: C32H29N5O2 Synonym: LY317615; 3-(1-Methyl-1H-indol-3-yl)-4-[1-[1-(2-pyridinylmethyl)-4-piperidinyl]-1H-indol-3-yl]-1H-pyrrole-2,5-dione