accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONFBr

CAS RN 1704065-18-8

IUPAC Name: 4-[(2-bromo-5-fluorophenyl)methyl]morpholine
Synonyms: 4-[(2-bromo-5-fluorophenyl)methyl]morpholine
Molecular Weight (g/mol): 274.13
Molecular Formula: C11H13BrFNO
InChi Key: IKTRFXHEKDRRIU-UHFFFAOYSA-N
SMILES: FC1=CC(CN2CCOCC2)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results