accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1704740-52-2

IUPAC Name: N-(4-{[3-cyano-1-(2-methylpropyl)-1H-indol-5-yl]oxy}phenyl)pyrrolidine-2-carboxamide
Synonyms: N-(4-{[3-cyano-1-(2-methylpropyl)indol-5-yl]oxy}phenyl)pyrrolidine-2-carboxamide
Molecular Weight (g/mol): 402.5
Molecular Formula: C24H26N4O2
InChi Key: OOGHGWKBJXQNEJ-UHFFFAOYNA-N
SMILES: CC(C)CN1C=C(C#N)C2=CC(OC3=CC=C(NC(=O)C4CCCN4)C=C3)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results