accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 170569-86-5

IUPAC Name: 4-[5-(4-chlorophenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzene-1-sulfonamide
Synonyms: 4-[5-(4-chlorophenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
Molecular Weight (g/mol): 401.79
Molecular Formula: C16H11ClF3N3O2S
InChi Key: NSQNZEUFHPTJME-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(C=C1)N1N=C(C=C1C1=CC=C(Cl)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results