accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONN

CAS RN 170632-47-0

IUPAC Name: [5-(1-benzyl-1H-indazol-3-yl)furan-2-yl]methanol
Synonyms: lificiguat
Molecular Weight (g/mol): 304.35
Molecular Formula: C19H16N2O2
InChi Key: OQQVFCKUDYMWGV-UHFFFAOYSA-N
SMILES: OCC1=CC=C(O1)C1=NN(CC2=CC=CC=C2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results