accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1706462-77-2

IUPAC Name: 2-oxa-5-azaspiro[3.5]nonane; oxalic acid
Synonyms: 2-oxa-5-azaspiro[3.5]nonane; oxalic acid
Molecular Weight (g/mol): 217.22
Molecular Formula: C9H15NO5
InChi Key: OQWJBTNQYYQHRU-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.C1OCC11CCCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results