accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 170727-02-3

IUPAC Name: 3-bromothiophene-2-sulfonyl chloride
Synonyms: 3-bromothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 261.53
Molecular Formula: C4H2BrClO2S2
InChi Key: OPLMNCCLQDVCTP-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=C(Br)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results