accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONOOH3CH3CCH3

CAS RN 170844-49-2

IUPAC Name: 1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate
Synonyms: 1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate
Molecular Weight (g/mol): 243.3
Molecular Formula: C12H21NO4
InChi Key: PKDQVUYUWUHNMG-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results