accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONBr

CAS RN 17117-17-8

IUPAC Name: 3-bromo-5-ethoxypyridine
Synonyms: 3-bromo-5-ethoxypyridine
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: FYUMRYNAZSTXJV-UHFFFAOYSA-N
SMILES: CCOC1=CC(Br)=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results