Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-ethoxypyridine 97.0+%, TCI America™
SDP

Catalog No. B50005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50005G 5 g
B50001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50005G Supplier TCI America Supplier No. B50005G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17117-17-8
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD00234116
InChI Key FYUMRYNAZSTXJV-UHFFFAOYSA-N
PubChem CID 12293480
IUPAC Name 3-bromo-5-ethoxypyridine
SMILES CCOC1=CC(=CN=C1)Br

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 240°C
Quantity 5 g
Formula Weight 202.05
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-ethoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.