CAS RN 171259-81-7
IUPAC Name:
2-amino-3-(5-methyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid hydrobromide
Synonyms:
AMPA hydrobromide
Molecular Weight (g/mol):
267.08
Molecular Formula:
C7H11BrN2O4
InChi Key:
KUAHVIUZGLGASU-UHFFFAOYNA-N
SMILES:
Br.CC1=C(CC(N)C(O)=O)C(=O)NO1
1
–
1
of
1
results
1
R&D Systems™ (RS)-AMPA hydrobromide
CAS: 171259-81-7 Molecular Formula: C7H11BrN2O4