accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1715-30-6

IUPAC Name: 1-{N''-[6-({amino[N''-(2-ethylhexyl)carbamimidamido]methylidene}amino)hexyl]carbamimidamido}-N'-(2-ethylhexyl)methanimidamide dihydrochloride
Synonyms: 1-{N''-[6-({amino[N''-(2-ethylhexyl)carbamimidamido]methylidene}amino)hexyl]carbamimidamido}-N'-(2-ethylhexyl)methanimidamide dihydrochloride
Molecular Weight (g/mol): 581.72
Molecular Formula: C26H58Cl2N10
InChi Key: BRJJFBHTDVWTCJ-UHFFFAOYNA-N
SMILES: Cl.Cl.CCCCC(CC)CN=C(N)NC(N)=NCCCCCCN=C(N)NC(N)=NCC(CC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results