Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Alexidine dihydrochloride
SDP

Catalog No. 397950 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-795-0 Supplier R&D Systems™ Supplier No. 3979/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of PTPMT1

Selective inhibitor of protein tyrosine phosphatases localized to mitochondrion 1 (PTPMT1) (IC50 = 1.08 μM in vitro). Stimulates increased insulin secretion by β-cells in rat pancreatic islets. Displays antitcancer properties in FaDu cells. Also preserves functional hematopoietic stem cells ex vivo.

Chemical Identifiers

CAS 1715-30-6
Molecular Formula C26H58Cl2N10

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 581.71
Purity >98%
Molecular Weight (g/mol) 581.72
Chemical Name or Material Alexidine dihydrochloride
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.