accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOOONHNNOO(Z)

CAS RN 1718-34-9

IUPAC Name: sodium (3Z)-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Synonyms: alizarine yellow R
Molecular Weight (g/mol): 309.21
Molecular Formula: C13H8N3NaO5
InChi Key: HKSANXZSKDUDBD-SOCRLDLMSA-M
SMILES: [Na+].[O-]C(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (2)
  • (2)
Grade 
  • (1)
  • (2)
Color 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)