Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Alizarin Yellow R sodium salt, 96%, pure

Catalog No. AC400490250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC400490250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC400490250 Supplier Thermo Scientific Chemicals Supplier No. 400490250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1718-34-9 | C13H8N3NaO5 | 309.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1718-34-9
Molecular Formula C13H8N3NaO5
Molecular Weight (g/mol) 309.213
MDL Number MFCD00067121
InChI Key HKSANXZSKDUDBD-UHFFFAOYSA-M
Synonym 5-(4-Nitrophenylazo)-salicylic acid sodium salt, Mordant Yellow 3R, 2-Hydroxy-5-((4-nitrophenyl)azo)benzoic acid monosodium salt
PubChem CID 44135758
IUPAC Name sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Loss on Drying 5% max. (105°C)
Packaging Glass bottle
Assay Percent Range 96%
UV_Vis 95% min.
Visual Transition Interval from pH 10.2 (yellow) to pH 12.0 (red)
Color Brown to Orange
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 309.2
Grade Pure
Chemical Name or Material Alizarin Yellow R sodium salt, 96%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.