accessibility menu, dialog, popup

UserName

Viewing profile as user:
2H2H2H2H2H2H2H2H2H2H

CAS RN 1719-06-8

IUPAC Name: (²H₁₀)anthracene
Synonyms: (²H₁₀)anthracene
Molecular Weight (g/mol): 188.3
Molecular Formula: C14H10
InChi Key: MWPLVEDNUUSJAV-LHNTUAQVSA-N
SMILES: [2H]C1=C([2H])C2=C([2H])C3=C([2H])C([2H])=C([2H])C([2H])=C3C([2H])=C2C([2H])=C1[2H]
Element 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)