accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 172035-86-8

IUPAC Name: 3-(iodomagnesio)thiophene
Synonyms: 3-(iodomagnesio)thiophene
Molecular Weight (g/mol): 234.34
Molecular Formula: C4H3IMgS
InChi Key: LDERXYCTDNLPTD-UHFFFAOYSA-M
SMILES: I[Mg]C1=CSC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results