Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Thienylmagnesium iodide, 0.3M solution in THF, AcroSeal™

Catalog No. AC433650500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433650500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433650500 Supplier Thermo Scientific Chemicals Supplier No. 433650500
Add to Cart

CAS: 172035-86-8 | C4H3IMgS | 234.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172035-86-8 , 109-99-9
Molecular Formula C4H3IMgS
Molecular Weight (g/mol) 234.34
MDL Number MFCD03427252
InChI Key LDERXYCTDNLPTD-UHFFFAOYSA-M
Synonym 3-thienylmagnesium iodide, magnesium, iodo-3-thienyl, iodo 3-thienyl magnesium, 3-thienyl magnesium iodide, thiophen-3-ylmagnesium iodide, 3-iodomagnesio thiophene, lderxyctdnlptd-uhfffaoysa-m, magnesium,3h-thiophen-3-ide,iodide, 3-thienylmagnesium iodide 0.5 m in tetrahydrofuran, 3-thienylmagnesium iodide solution, 0.3 m in thf
PubChem CID 10752186
IUPAC Name 3-(iodomagnesio)thiophene
SMILES I[Mg]C1=CSC=C1

Specifications

Boiling Point 65.0°C
Color Yellow to Brown
CAS Min % 5.19, 89.63
CAS Max % 10.37, 94.82
Physical Form Liquid
Quantity 50 mL
Flash Point −17°C
Solubility Information Solubility in water: reacts
Formula Weight 234.34
Concentration or Composition (by Analyte or Components) 0.20 to 0.40M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.904
Density 0.9040g/mL
Chemical Name or Material 3-Thienylmagnesium iodide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Suspected of causing cancer if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
May form explosive per
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.