accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17213-58-0

H3COOCH3H2NO

CAS RN 17213-58-0

IUPAC Name: 3,5-dimethoxybenzamide
Synonyms: 3,5-dimethoxybenzamide
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: YTLRWVNYANKXOW-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)C(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results