accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CH3NH3CH3C

CAS RN 1722-26-5

IUPAC Name: triethylamine borane
Synonyms: triethylamine borane
Molecular Weight (g/mol): 115.03
Molecular Formula: C6H18BN
InChi Key: KDKNBGWKECQREO-UHFFFAOYSA-N
SMILES: B.CCN(CC)CC
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)