accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOH3C(E/Z)

CAS RN 17224-17-8

IUPAC Name: 1-(4-{[(4-ethoxyphenyl)methylidene]amino}phenyl)ethan-1-one
Synonyms: 1-(4-{[(4-ethoxyphenyl)methylidene]amino}phenyl)ethanone
Molecular Weight (g/mol): 267.33
Molecular Formula: C17H17NO2
InChi Key: HCDYNBNSFSMUIM-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=NC2=CC=C(C=C2)C(C)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results