CAS RN 172478-00-1
CAS RN 172478-00-1
IUPAC Name:
tert-butyl N-methyl-N-(pyrrolidin-3-yl)carbamate
Synonyms:
tert-butyl N-methyl-N-(pyrrolidin-3-yl)carbamate
Molecular Weight (g/mol):
200.28
Molecular Formula:
C10H20N2O2
InChi Key:
XYKYUXYNQDXZTD-UHFFFAOYNA-N
SMILES:
CN(C1CCNC1)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine 96.0+%, TCI America™
CAS: 172478-00-1 Molecular Formula: C10H20N2O2 Molecular Weight (g/mol): 200.282 MDL Number: MFCD00191665 InChI Key: XYKYUXYNQDXZTD-UHFFFAOYSA-N Synonym: 3-(N-Boc-N-methylamino)pyrrolidine PubChem CID: 2761404 IUPAC Name: tert-butyl N-methyl-N-pyrrolidin-3-ylcarbamate SMILES: CC(C)(C)OC(=O)N(C)C1CCNC1