accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 172611-72-2

CH3H3COOOH3CCH3

CAS RN 172611-72-2

IUPAC Name: 5,5-dimethyl-2-(3-methylbutanoyl)cyclohexane-1,3-dione
Synonyms: 5,5-dimethyl-2-(3-methylbutanoyl)cyclohexane-1,3-dione
Molecular Weight (g/mol): 224.3
Molecular Formula: C13H20O3
InChi Key: HNPWTDUZIXAJSA-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)C1C(=O)CC(C)(C)CC1=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)