accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3O

CAS RN 1728-46-7

IUPAC Name: 2-tert-butylcyclohexan-1-one
Synonyms: 2-tert-butylcyclohexanone
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: ZRYDPLOWJSFQAE-UHFFFAOYNA-N
SMILES: CC(C)(C)C1CCCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results