CAS RN 172806-20-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Guanosine 3',5'-cyclic Monophosphorothioate, β-Phenyl-1,N2-etheno-8-bromo-, Rp-Isomer, Sodium Salt
A metabolically-stable, competitive inhibitor of protein kinase G types 1α and 1β (Ki = 30 nM).