CAS RN 172895-15-7
IUPAC Name:
1-[(3,4-dimethoxyphenyl)methyl]-6-methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole hydrochloride
Synonyms:
1-[(3,4-dimethoxyphenyl)methyl]-6-methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole hydrochloride
Molecular Weight (g/mol):
372.89
Molecular Formula:
C21H25ClN2O2
InChi Key:
BKAZOTIBKRWLQA-UHFFFAOYNA-N
SMILES:
Cl.COC1=C(OC)C=C(CC2NCCC3=C2NC2=CC=C(C)C=C32)C=C1
1
–
1
of
1
results
1
R&D Systems™ LY 272015 hydrochloride
CAS: 172895-15-7 Molecular Formula: C21H25ClN2O2