CAS RN 172895-39-5
IUPAC Name:
1-[(2-chloro-3,4-dimethoxyphenyl)methyl]-6-methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole hydrochloride
Synonyms:
1-[(2-chloro-3,4-dimethoxyphenyl)methyl]-6-methyl-1H,2H,3H,4H,9H-pyrido[3,4-b]indole hydrochloride
Molecular Weight (g/mol):
407.34
Molecular Formula:
C21H24Cl2N2O2
InChi Key:
KPXKZZURYAXZQE-UHFFFAOYNA-N
SMILES:
Cl.COC1=C(OC)C(Cl)=C(CC2NCCC3=C2NC2=CC=C(C)C=C32)C=C1
1
–
1
of
1
results
1
R&D Systems™ LY 266097 hydrochloride
CAS: 172895-39-5 Molecular Formula: C21H24Cl2N2O2