accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 1731-92-6

IUPAC Name: methyl heptadecanoate
Synonyms: heptadecanoic acid, methyl ester
Molecular Weight (g/mol): 284.48
Molecular Formula: C18H36O2
InChi Key: HUEBIMLTDXKIPR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)