Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl margarate, EMD-Millipore

Catalog No. M1097540005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1097540005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1097540005 Supplier MilliporeSigma Supplier No. 1.09754.0005

missing translation for 'validMainframeMsg'

Fatty acid methyl esters

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1731-92-6
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.484
InChI Key HUEBIMLTDXKIPR-UHFFFAOYSA-N
Synonym Methyl heptadecanate
PubChem CID 15609
IUPAC Name methyl heptadecanoate
SMILES CCCCCCCCCCCCCCCCC(=O)OC

Specifications

Packaging Glass vials
Density Density: 0.78g/cm3 (20°)
Assay Percent Range ≥99.0%
Quantity 5 mL
Formula Weight 284.48
Chemical Name or Material Methyl margarate
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.